Merck
  • Acute illness associated with use of pest strips - seven U.S. States and Canada, 2000-2013.
Acute illness associated with use of pest strips - seven U.S. States and Canada, 2000-2013.
MMWR. Morbidity and mortality weekly report (2014-01-17)
Rebecca J Tsai, Jennifer Sievert, Joanne Prado, Kaci Buhl, Dave L Stone, Mathias Forrester, Shelia Higgins, Yvette Mitchell, Abby Schwartz, Geoffrey M Calvert
ABSTRACT
Dichlorvos-impregnated resin strips (DDVP pest strips) are among the few organophosphate products still available for indoor residential use. The residential uses for most other organophosphate products, including most DDVP products, were canceled because they posed unreasonable risks to children. DDVP pest strips act by inhibiting acetylcholinesterase activity in the brain and nerves of insect pests and are designed to gradually release DDVP vapor for up to 4 months. Acute illnesses in humans associated with nonlethal acute exposures usually resolve completely, but recovery is not always rapid. To assess the frequency of acute illnesses associated with DDVP pest strips, cases from 2000 through June 2013 were sought from the 12 states that participate in the Sentinel Event Notification System for Occupational Risks (SENSOR)-Pesticides Program, the National Pesticide Information Center (NPIC), and Health Canada.* A total of 31 acute DDVP pest strip-related illness cases were identified in seven U.S. states and Canada. The majority of these illnesses resulted from use of the product in commonly occupied living areas (e.g., kitchens and bedrooms), in violation of label directions. Although 26 of the 31 cases involved mild health effects of short duration, five persons had moderate health effects. Illnesses caused by excess exposure to DDVP pest strips can be reduced by educating the public about the proper usage of DDVP pest strips and with improvements in label directions.
Product Description
Supelco
Dichlorvos, PESTANAL®, analytical standard
Pricing and availability is not currently available.