Merck
  • Effects of pH on the production of phosphate and pyrophosphate by matrix vesicles' biomimetics.
Effects of pH on the production of phosphate and pyrophosphate by matrix vesicles' biomimetics.
Calcified tissue international (2013-08-15)
Ana Maria S Simão, Maytê Bolean, Marc F Hoylaerts, José Luis Millán, Pietro Ciancaglini
ABSTRACT
During endochondral bone formation, chondrocytes and osteoblasts synthesize and mineralize the extracellular matrix through a process that initiates within matrix vesicles (MVs) and ends with bone mineral propagation onto the collagenous scaffold. pH gradients have been identified in the growth plate of long bones, but how pH changes affect the initiation of skeletal mineralization is not known. Tissue-nonspecific alkaline phosphatase (TNAP) degrades extracellular inorganic pyrophosphate (PPi), a mineralization inhibitor produced by ectonucleotide pyrophosphatase/phosphodiesterase-1 (NPP1), while contributing Pi from ATP to initiate mineralization. TNAP and NPP1, alone or combined, were reconstituted in dipalmitoylphosphatidylcholine liposomes to mimic the microenvironment of MVs. The hydrolysis of ATP, ADP, AMP, and PPi was studied at pH 8 and 9 and compared to the data determined at pH 7.4. While catalytic efficiencies in general were higher at alkaline pH, PPi hydrolysis was maximal at pH 8 and indicated a preferential utilization of PPi over ATP at pH 8 versus 9. In addition, all proteoliposomes induced mineral formation when incubated in a synthetic cartilage lymph containing 1 mM ATP as substrate and amorphous calcium phosphate or calcium-phosphate-phosphatidylserine complexes as nucleators. Propagation of mineralization was significantly more efficient at pH 7.5 and 8 than at pH 9. Since a slight pH elevation from 7.4 to 8 promotes considerably more hydrolysis of ATP, ADP, and AMP primarily by TNAP, this small pH change facilitates mineralization, especially via upregulated PPi hydrolysis by both NPP1 and TNAP, further elevating the Pi/PPi ratio, thus enhancing bone mineralization.
Sigma-Aldrich
Brij® L23 solution, 30 % (w/v) in H2O
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
1,2-Dipalmitoyl-rac-glycero-3-phosphocholine, ~99%
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Phosphodiesterase I from Crotalus atrox (Western Diamondback Rattlesnake), Type IV, crude dried venom
Sigma-Aldrich
Decaethylene glycol mono­dodecyl ether, nonionic surfactant
Sigma-Aldrich
Thesit®, for membrane research
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
ECO Brij® L23
Sigma-Aldrich
ECO BRIJ® L4, average Mn ~362
Sigma-Aldrich
Phosphodiesterase I from Crotalus adamanteus venom, Type VI, crude dried venom
Sigma-Aldrich
Brij® L23, suitable for Stein-Moore chromatography
Sigma-Aldrich
Brij® L4, average Mn ~362
Sigma-Aldrich
Phosphodiesterase I from Crotalus adamanteus venom, vial of ≥100 units, Purified
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether