Merck
  • Biocomposite scaffolds containing chitosan/alginate/nano-silica for bone tissue engineering.
Biocomposite scaffolds containing chitosan/alginate/nano-silica for bone tissue engineering.
Colloids and surfaces. B, Biointerfaces (2013-05-15)
J A Sowjanya, J Singh, T Mohita, S Sarvanan, A Moorthi, N Srinivasan, N Selvamurugan
ABSTRACT
Bone tissue engineering is a promising alternative method for treating bone loss by a combination of biomaterials and cells. In this study, we fabricated biocomposite scaffolds by blending chitosan (CS), alginate (Alg) and nano-silica (nSiO2), followed by freeze drying. The prepared scaffolds (CS/Alg, CS/Alg/nSiO2) were characterized by SEM, FT-IR and XRD analyses. In vitro studies such as swelling, biodegradation, biomineralization, protein adsorption and cytotoxicity were also carried out. The scaffolds possessed a well-defined porous architecture with pore sizes varying from 20 to 100 μm suitable for cell infiltration. The presence of nSiO2 in the scaffolds facilitated increased protein adsorption and controlled swelling ability. The scaffolds were biodegradable and the addition of nSiO2 improved apatite deposition on these scaffolds. There was no significant cytotoxicity effect of these CS/Alg/nSiO2 scaffolds towards osteolineage cells. Thus, these results indicate that CS/Alg/nSiO2 scaffolds may have potential applications for bone tissue engineering.
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity