Merck
  • Tetraaqua(3,4,7,8-tetramethyl-1,10-phenanthroline-kappa2N,N')zinc(II) thiosulfate.
Tetraaqua(3,4,7,8-tetramethyl-1,10-phenanthroline-kappa2N,N')zinc(II) thiosulfate.
Acta crystallographica. Section C, Crystal structure communications (2004-09-04)
M Enriqueta Díaz de Vivar, Sergio Baggio, Andrés Ibañez, Ricardo Baggio
ABSTRACT
In the title complex of zinc(II) with 3,4,7,8-tetramethyl-1,10-phenanthroline (tmph), viz. [Zn(C16H16N2)(H2O)4](S2O3), the metal atom has a monomeric octahedral ZnN2O4 complex environment comprising two N-atom donors from the tmph group and four aqua O-atom donors. The complex cation is connected to four thiosulfate anions through a compact hydrogen-bonding network involving all coordinated aqua H-atom donors and all the outer acceptors (O and S) of the anion.
Sigma-Aldrich
3,4,7,8-Tetramethyl-1,10-phenanthroline, ≥98%