Merck
  • A Bcl-xL-Drp1 complex regulates synaptic vesicle membrane dynamics during endocytosis.
A Bcl-xL-Drp1 complex regulates synaptic vesicle membrane dynamics during endocytosis.
Nature cell biology (2013-06-25)
Hongmei Li, Kambiz N Alavian, Emma Lazrove, Nabil Mehta, Adrienne Jones, Ping Zhang, Pawel Licznerski, Morven Graham, Takuma Uo, Junhua Guo, Christoph Rahner, Ronald S Duman, Richard S Morrison, Elizabeth A Jonas
ABSTRACT
Following exocytosis, the rate of recovery of neurotransmitter release is determined by vesicle retrieval from the plasma membrane and by recruitment of vesicles from reserve pools within the synapse, which is dependent on mitochondrial ATP. The anti-apoptotic Bcl-2 family protein Bcl-xL also regulates neurotransmitter release and recovery in part by increasing ATP availability from mitochondria. We now find, that Bcl-xL directly regulates endocytic vesicle retrieval in hippocampal neurons through protein-protein interaction with components of the clathrin complex. Our evidence suggests that, during synaptic stimulation, Bcl-xL translocates to clathrin-coated pits in a calmodulin-dependent manner and forms a complex with the GTPase Drp1, Mff and clathrin. Depletion of Drp1 produces misformed endocytic vesicles. Mutagenesis studies suggest that formation of the Bcl-xL-Drp1 complex is necessary for the enhanced rate of vesicle endocytosis produced by Bcl-xL, thus providing a mechanism for presynaptic plasticity.
Sigma-Aldrich
Mitochondria/Cytosol Fractionation Kit, This Mitochondria/Cytosol Fractionation Kit provides reagents for quick & efficient isolation of intact mitochondria from cultured cells.