Merck
  • Temperature sensitive dopamine-imprinted (N,N-methylene-bis-acrylamide cross-linked) polymer and its potential application to the selective extraction of adrenergic drugs from urine.
Temperature sensitive dopamine-imprinted (N,N-methylene-bis-acrylamide cross-linked) polymer and its potential application to the selective extraction of adrenergic drugs from urine.
Journal of chromatography. A (2006-03-15)
Roongnapa Suedee, Vatcharee Seechamnanturakit, Bhutorn Canyuk, Chitchamai Ovatlarnporn, Gary P Martin
ABSTRACT
A temperature sensitive dopamine-imprinted polymer was prepared in 80% aqueous methanol solution by free-radical cross-linking co-polymerisation of methacrylic acid and acrylamide at 60 degrees C in the presence of N,N-methylene-bis-acrylamide as the cross-linker and dopamine hydrochloride as template molecule. The resulting molecularly imprinted polymer (MIP) formed temperature responsive materials, which could be used for the selective separation of appropriate dopamine and adrenergic compounds from a liquid matrix at ambient temperatures. The thermoresponsive MIP exhibited a swelling-deswelling transition in 80% aqueous methanol solution at about 35 degrees C. The capacity of the thermoresponsive MIP to recognise the template molecule when present in aqueous methanol solution changed with temperature, with the highest selectivity found at 35 degrees C. Additionally, binding parameters obtained from Scatchard analyses indicate that increasing temperature resulted in an increased affinity and binding capacity of specific binding sites, but had less effect on non-selective binding sites. Subsequently, the thermoresponsive MIP was tested for its application as a sorbent material, utilisable in the selective solid-phase extraction (SPE) of dopamine and other adrenergic compounds (epinephrine, isoproterenol, salbutamol and serotonin) from urine samples. It was shown that the compounds that were structurally related to dopamine could be removed by elution, while dopamine and serotonin, the analytes of interest, remained strongly adsorbed to the adsorbent during SPE applications. The thermoresponsive MIP displayed different efficiency in clean-up and enrichments using the SPE protocol at different temperatures.
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Acrylamide:N,N′-Methylenebisacrylamide 29:1, BioXtra, for molecular biology