Merck
  • A triptycene-based polymer of intrinsic microposity that displays enhanced surface area and hydrogen adsorption.
A triptycene-based polymer of intrinsic microposity that displays enhanced surface area and hydrogen adsorption.
Chemical communications (Cambridge, England) (2007-02-07)
Bader S Ghanem, Kadhum J Msayib, Neil B McKeown, Kenneth D M Harris, Zhigang Pan, Peter M Budd, Anna Butler, James Selbie, David Book, Allan Walton
ABSTRACT
A novel triptycene-based polymer of intrinsic microporosity (Trip-PIM) displays enhanced surface area (1065 m2 g(-1)) and reversibly adsorbs 1.65% hydrogen by mass at 1 bar/77 K and 2.71% at 10 bar/77 K.
Sigma-Aldrich
Triptycene, 98%