Merck
  • Human N-demethylation of (S)-mephenytoin by cytochrome P450s 2C9 and 2B6.
Human N-demethylation of (S)-mephenytoin by cytochrome P450s 2C9 and 2B6.
Drug metabolism and disposition: the biological fate of chemicals (1998-08-11)
J W Ko, Z Desta, D A Flockhart
ABSTRACT
We tested the ability of human liver microsomes (HLMs) and recombinant human cytochrome P450 (CYP or P450) isoforms to catalyze the N-demethylation of nirvanol-free (S)-mephenytoin [(S)-MP] in vitro. In mixed HLMs, the kinetics of (S)-MP N-demethylation suggested two contributing activities. A high-affinity/low-capacity component exhibited a KM of 174.1 microM and a Vmax of 170.5 pmol/mg protein/min, whereas a low-affinity/high-capacity component exhibited a KM of 1911 microM and a Vmax of 3984 pmol/mg protein/min. The activity of the high-affinity component was completely abolished by sulfaphenazole, with little effect on the low-affinity component. Of the recombinant P450 isoforms tested, only CYP2B6 and CYP2C9 formed nirvanol from (S)-MP. The KM value (150 +/- 42 microM) derived for recombinant CYP2C9 was close to that obtained for the high-affinity/low-capacity component in mixed HLMs (KM = 174.1 microM). The predicted contribution of this activity at concentrations (1-25 microM) achieved after a single 100-mg dose of racemic MP is approximately 30% of the rate of nirvanol formation. At concentrations of >1000 microM, we estimate that >90% of the rate can be explained by the low-affinity activity (CYP2B6). Therefore, the N-demethylation of (S)-MP to nirvanol may be a useful means of probing the activity of CYP2B6 in vitro when concentrations of >1000 microM are used, but it is unlikely to be a suitable phenotyping tool for this isoform in vivo, where concentrations of >1000 microM are rarely encountered.
Sigma-Aldrich
(S)-(+)-Nirvanol
Sigma-Aldrich
(R)-(−)-Nirvanol