- Purification and properties of beta-N-Acetylhexosaminidase from cabbage.
Purification and properties of beta-N-Acetylhexosaminidase from cabbage.
beta-N-Acetylhexosaminidase was purified from the extract of cabbage by sequential steps of ammonium sulfate fractionation, chromatofocusing, DEAE-Sepharose CL-6B ion exchange chromatography and Sephacryl S-200 HR gel filtration. By these steps, the purity of the enzyme increased by 256 fold with a recovery of 8%. The purified enzyme was homogeneous as examined by native PAGE. It showed an optimal pH of 4, an optimal temperature of 60 degrees C and a Km of 0.94 mM for hydrolysis of pNp-beta-GlcNAc. The molecular mass of the enzyme determined from filtration through Sephacryl S-200 was 150 kDa. Three subunits with molecular mass of 64, 57 and 51 kDa were observed as determined by SDS-PAGE. NBS (0.025 mM), DEPC (3 mM) and WRK (30 mM) significantly inhibited the activity of the enzyme. The enzyme also showed activity toward pNp-beta-GalNAc, N,N'-diacetylchitobiose, N,N',N"-triacetylchitotriose and N,N',N",N"'-tetraacetyl chitotetraose but showed no activity toward pNp-alpha-GlcNAc, chitin and ethylene glycol chitin.