Merck
  • Synthesis of [35S]thiophosphoryl adenylic acid, utilizing a general procedure for [35S]thiophosphoryl chloride production.
Synthesis of [35S]thiophosphoryl adenylic acid, utilizing a general procedure for [35S]thiophosphoryl chloride production.
Analytical biochemistry (1993-02-15)
J T Slama, A M Simmons, T M Hernandez, R W Keenan
ABSTRACT
Elemental [35S]sulfur was shown to equilibrate with the sulfur of thiophosphoryl chloride when these materials are heated together. This isotopic exchange reaction is the basis of a convenient, microscale synthesis of high specific activity [35S]PSCl3. [35S]Thiophosphoryl chloride is otherwise not commercially available except through custom synthesis. The labeled thiophosphoryl chloride was used in a novel procedure for the preparation of [35S]adenosine 5'-phosphorothioate. This isotopic exchange method should find wide application in the synthesis of many radiolabeled thiophosphoryl esters which utilize PSCl3 as the source of the thiophosphoryl group.
Sigma-Aldrich
Thiophosphoryl chloride, 98%