Merck
  • Virus-induced airway inflammation and hyperresponsiveness in the guinea-pig is inhibited by levodropropizine.
Virus-induced airway inflammation and hyperresponsiveness in the guinea-pig is inhibited by levodropropizine.
Naunyn-Schmiedeberg's archives of pharmacology (1993-08-01)
G Folkerts, H J van der Linde, C Omini, F P Nijkamp
ABSTRACT
Intratracheal Parainfluenza type 3 (PI-3) virus inoculation of guinea pigs leads to a non-specific airway hyperresponsiveness in vivo and in vitro which coincides with a significant increase in the number of inflammatory cells in the broncho-alveolar lavage fluid (90% increase, 4 days after inoculation). The activity of the bronchoalveolar cells, as measured by the chemiluminescence production of infected animals is significantly diminished (34.2%, 4 days after inoculation) after renewed stimulation with PI-3 virus in vitro as compared to the chemiluminescence production by bronchoalveolar cells obtained from control guinea pigs. Pretreatment of the guinea-pigs with the antitussive agent levodropropizine, administered intra-peritoneally twice a day for five successive days at a dose of 10 mg/kg, prevents the virus-induced airway hyperresponsiveness in vivo and in vitro, and inhibits the influx of broncho-alveolar cells. Levodropropizine at a dose of 1 mg/kg did not modulate these responses. Further, the decrease in chemiluminescence production of broncho-alveolar cells obtained from virus-infected animals after PI-3 virus stimulation in vitro was inhibited by levodropropizine (10 mg/kg). These data demonstrate the ability of levodropropizine to counteract the hyperresponsiveness phenomenon and the associated inflammatory event induced by PI-3 virus, an effect which may be due to its capacity to act on the peptidergic system or may be due to the anti-allergic/bronchoconstrictor property of this compound.
Product Description
Supelco
(±)-Dropropizine, analytical standard, for drug analysis
Levodropropizine, European Pharmacopoeia (EP) Reference Standard