Merck
Antitussive effects of levodropropizine in the dog.
Arzneimittel-Forschung (1994-02-01)
P L Munt, G Clavenna, D R Algate, R M Leach
ABSTRACT
The antitussive activity of levodropropizine (S(-)3-(4-phenyl-piperazine-1-yl)-propane-1,2-diol, DF 526, CAS 99291-25-5) was evaluated after oral administration to the conscious dog. Levodropropizine had a good antitussive activity, comparable with, but having a longer duration of action than dropropizine, the racemate from which it is derived. The antitussive activity of levodropropizine in the dog was approximately 1/20 of that of codeine phosphate.
Product Description
Supelco
(±)-Dropropizine, analytical standard, for drug analysis
Levodropropizine, European Pharmacopoeia (EP) Reference Standard