Merck
  • Rapid detection of sublethally impaired cells of Enterobacteriaceae in dried foods.
Rapid detection of sublethally impaired cells of Enterobacteriaceae in dried foods.
Applied microbiology (1970-08-01)
D A Mossel, M A Ratto
ABSTRACT
Samples of dried foods, feeds, and drugs were examined for Enterobacteriaceae by using buffered glucose-Brilliant Green-bile broth as the final enrichment medium and deep tubes of violet-red-bile-glucose-agar for confirmation. The pre-enrichment treatments used for the resuscitation of sublethally impaired cells were overnight incubation in lactose broth versus incubation for 1 to 6 hr at room temperature in shallow layers of tryptone soya peptone broth. The latter restoration treatment appeared to be significantly more productive.
Product Description
Millipore
Violet Red Bile Glucose Agar, suitable for microbiology, NutriSelect® Plus