Merck
  • Interaction of non-conjugated olefinic substrate analogues with dopamine beta-monooxygenase: catalysis and mechanism-based inhibition.
Interaction of non-conjugated olefinic substrate analogues with dopamine beta-monooxygenase: catalysis and mechanism-based inhibition.
The Biochemical journal (1995-02-15)
S R Sirimanne, S W May
ABSTRACT
The reaction of dopamine beta-monooxygenase (DBM; EC 1.14.17.1) with the prototypical non-conjugated olefinic substrate, 2-(1-cyclohexenyl)ethylamine (CyHEA) [see Sirimanne and May (1988) J. Am. Chem. Soc. 110, 7560-7561], was characterized. CyHEA undergoes facile DBM-catalysed allylic hydroxylation to form (R)-2-amino-1-(1-cyclohexenyl)ethanol (CyHEA-OH) without detectable epoxidation or allylic hydroxylation to form (R)-2-amino-1-(1-cyclohexenyl)ethanol (CyHEA-OH) without detectable epoxidation or allylic rearrangement, and with stereochemistry consistent with that of DBM-catalysed benzylic hydroxylation and sulphoxidation. The kcat. of 90 s-1 for CyHEA oxygenation is about 75% of the kcat. for tyramine, the substrate commonly used in assays of DBM activity. DBM-catalysed oxygenation of CyHEA also results in mechanism-based inactivation of DBM, with the inactivation reaction yielding kinact. = 0.3 min-1 at pH 5.0 and 37 degrees C, and a partition ratio of 16,000. Although both CyHEA turnover and inactivation exhibit normal kinetics, CyHEA processing also results in gradual depletion of copper from DBM; however, mechanism-based irreversible DBM inactivation occurs independent of this copper depletion when sufficient copper is present in the assay solution. A likely mechanism for turnover-dependent DBM inactivation by CyHEA involves initial abstraction of an allylic hydrogen to form a resonance-stabilized allylic radical, which can then either partition to product or undergo attack by an active-site residue. Acyclic, non-conjugated olefinic analogues exhibit diminished substrate activity toward DBM. Thus, kcat. for oxygenation of cis-2-hexenylamine, which also produces only allylic alcohol product, is only 14% of that for CyHEA. Similarly, kinact./KI for turnover-dependent inactivation by the acyclic olefin 2-aminomethyl-1-pentene is more than an order of magnitude smaller than that for benzylic olefins. Our results establish that DBM catalyses allylic oxygenation of a number of non-conjugated olefinic substrate analogues with neither epoxidation nor allylic rearrangement occurring. The absence of epoxide products from non-conjugated olefinic substrates implies an inability of the activated copper-oxygen species of DBM to effect radical cation formation from a non-conjugated olefinic moiety. The striking contrast between DBM and cytochrome P-450, which carries out both epoxidation and allylic oxidation with non-conjugated olefinic substrates, is probably a reflection of the differences in redox potential of the activated oxygen species operative for these two enzymes.
Sigma-Aldrich
2-(1-Cyclohexenyl)ethylamine, 97%