Merck
  • Effect of model inducers on cytochrome P450 activities of human hepatocytes in primary culture.
Effect of model inducers on cytochrome P450 activities of human hepatocytes in primary culture.
Drug metabolism and disposition: the biological fate of chemicals (1995-05-01)
M T Donato, J V Castell, M J Gómez-Lechón
ABSTRACT
The dealkylations of 7-ethoxy- and 7-pentoxyresorufin,p-nitrophenol hydroxylation, and regio- and stereoselective hydroxylation of testosterone were measured to study the stability and inducibility of cytochrome P450 activities in cultured human hepatocytes. The results showed that human hepatocytes in primary culture retain the ability to increase specific cytochrome P450 activities upon incubation with inducers. 3-Methylcholanthrene produced a strong increase (6- to 21-fold over control) in 7-ethoxyresorufin O-deethylase activity and a small enhancement (1.5- to 2.5-fold) of the p-nitrophenol hydroxylation rate. Incubation of cells with phenobarbital resulted in moderate increases in 7-pentoxyresorufin O-depentylation (1.5- to 2-fold) and in testosterone hydroxylation at 16 alpha (1.5- to 4.5-fold) and 16 beta (1.3- to 4-fold) positions. Ethanol specifically increased p-nitrophenol hydroxylase activity (1.5- to 3.5-fold) and reduced 15 beta- and 6 beta-hydroxylations of testosterone. Treatment of hepatocytes with dexamethasone produced an increase of almost all the activities studied, with 6 beta- (2- to 3-fold) and 16 beta-hydroxytestosterone (1.4- to 2.4-fold) formation showing the greatest enhancement. Clofibric acid exposure resulted in 1.5- to 3-fold increases in 7-pentoxyresorufin O-depentylase and in testosterone 6 beta- and 2 beta-hydroxylase activities. Isosafrol selectively increased 7-ethoxyresorufin O-deethylase activity (2- to 3-fold), and it moderately reduced the other activities studied.
Sigma-Aldrich
Isosafrol, mixture of cis and trans