Merck
  • Coexpression of 5-HT2A and 5-HT4 receptors coupled to distinct signaling pathways in human intestinal muscle cells.
Coexpression of 5-HT2A and 5-HT4 receptors coupled to distinct signaling pathways in human intestinal muscle cells.
Gastroenterology (1995-12-01)
J F Kuemmerle, K S Murthy, J R Grider, D C Martin, G M Makhlouf
ABSTRACT
The type and function of 5-hydroxytryptamine (5HT) receptors on intestinal muscle cells in humans are not known. 5-HT receptors were characterized pharmacologically and by radioligand binding. Contraction, relaxation, inositol 1,4,5-triphosphate (IP3) and adenosine 3',5'-cyclic monophosphate (cAMP) formation, and 5-HT binding were measured in dispersed muscle cells and in cells in which only one receptor type was preserved by selective receptor protection. 5-HT binding was completely inhibited by 5-HT and partially by 5-HT2A (ketanserin), 5-HT4 (SDZ-205,557), and 5-HT1p (N-acetyl-5-hydroxytryptophyl-5-hydroxytryptophan amide; 5-HTP-DP) receptor antagonists. 5-HT caused contraction that was inhibited by ketanserin and augmented by SDZ-205,557 and 5-HTP-DP. In the presence of ketanserin, 5-HT caused relaxation of cholecystokinin-contracted cells that was inhibited by SDZ-205,557 and 5-HTP-DP. 5-HT increased IP3, which was inhibited by ketanserin, and cAMP, which was inhibited by SDZ-205,557 and 5-HTP-DP. In cells with only 5-HT2A receptors, 5-HT caused contraction only, and residual binding was inhibited by ketanserin. In cells with only 5-HT4/5-HT1p receptors, 5-HT caused only relaxation and residual binding was inhibited by SDZ-205,557 and 5-HTP-DP. 5-HT2A receptors mediating contraction and 5-HT4 receptors mediating relaxation coexist on human intestinal muscle cells. The 5-HT4 receptors are closely similar or identical to 5-HT1p receptors.
Sigma-Aldrich
SDZ-205,557 hydrochloride, solid