- Ethyl nitrite. The in vivo synthesis of a mutagenic substance from tobacco smoke and ethanol.
Ethyl nitrite. The in vivo synthesis of a mutagenic substance from tobacco smoke and ethanol.
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry (1981-01-01)
J C MacDonald, A Jonsson
PMID7315123
ABSTRACT
Gas chromatography--mass spectrometry demonstrates the presence of ethyl nitrite in the breath of volunteers subjects who smoke and drink concurrently. The mutagenic compound is quantitated at the ppb level by gas chromatography with the selective electron capture detector. The compound is not detected in cases of only drinking or only smoking.