Merck
  • Apoptosis induced by tungsten carbide-cobalt nanoparticles in JB6 cells involves ROS generation through both extrinsic and intrinsic apoptosis pathways.
Apoptosis induced by tungsten carbide-cobalt nanoparticles in JB6 cells involves ROS generation through both extrinsic and intrinsic apoptosis pathways.
International journal of oncology (2013-02-19)
Jinshun Zhao, Linda Bowman, Ruth Magaye, Stephen S Leonard, Vincent Castranova, Min Ding
ABSTRACT
In this study, apoptosis and related signaling induced by WC-Co nanoparticles were investigated in JB6 cells and rat lung macrophages. Electron spin resonance (ESR) and fluorescent staining indicated that both WC-Co nanoparticles and fine particles stimulated reactive oxygen species (ROS) generation. Catalase exhibited an inhibitory effect on WC-Co nanoparticle-induced ROS as well as mitochondrial membrane permeability damage. Further study indicated that WC-Co nanoparticles elicited higher cytotoxicity and apoptotic induction than fine particles. Western blot analysis showed activation of proapoptotic factors including Fas, Fas-associated protein with death domain (FADD), caspase 3, 8 and 9, BID and BAX. In addition, both cytochrome c and apoptosis-inducing factor (AIF) were upregulated and released from mitochondria to the cytoplasm. Our findings demonstrate that, on a mass basis, WC-Co nanoparticles exhibit higher cytotoxicity and apoptotic induction than fine particles. Apoptosis induced by WC-Co nanoparticles and fine particles involves both extrinsic and intrinsic apoptosis pathways.
Sigma-Aldrich
Tungsten(IV) carbide, powder, 2 μm, ≥99%