Merck
  • Novel near infra-red fluorescent pH sensors based on 1-aminoperylene bisimides covalently grafted onto poly(acryloylmorpholine).
Novel near infra-red fluorescent pH sensors based on 1-aminoperylene bisimides covalently grafted onto poly(acryloylmorpholine).
Chemical communications (Cambridge, England) (2013-02-08)
Daniel Aigner, Sergey M Borisov, Peter Petritsch, Ingo Klimant
ABSTRACT
Novel pH sensors relying on 1-aminoperylene bisimide dyes covalently grafted onto cross-linked poly(acryloylmorpholine) are presented. They feature fluorescence in the near infra-red range and a large Stokes shift (>90 nm).
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%