Merck
Appetite suppressants in pregnancy.
Neuro endocrinology letters (2013-01-29)
Eva Manakova, Tereza Kralova, Lucie Hubičková Heringová
ABSTRACT
Both, obesity as well as anorexia may be associated with infertility and other complications of pregnancy. Weight loss during pregnancy is therefore considered a risk factor. Weight loss and appetite suppressant are contraindicated during pregnancy, but the unintended exposure is probably not associated with higher risk. Our work was focused on trends in the appetite suppressants use in the Czech Republic and their embryotoxicity. The pregnancies exposed to various appetite suppressants were followed prospectively in the years 1997-2012. The study group was compared to the comparison group which enrolled pregnant women exposed to non-teratogenic drugs. Drugs used as appetite suppressants were sibutramine and phentermine. Number of calls for this type of exposure was rare till 2005. Their number started to increase until 2009. Later, number of calls decreased because both drugs were withdrawn from the market. This finding reflects increasing tendency for the weight control in the group of fertile women in the Czech Republic. In our study, we did not reveal differences in pregnancy outcomes between study and comparison groups. However, we should be aware of the increasing food supplements exposure, that could be used as alternative to the appetite suppressants. Their potential risk results from the limited or completely absent control of their origin. Some of them have probably only placebo effect, but some of them could represent the risk.
Product Description
Supelco
Phentermine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.