Merck
A tripeptide-like prolinamide-thiourea as an aldol reaction catalyst.
Organic & biomolecular chemistry (2012-06-22)
Stamatis Fotaras, Christoforos G Kokotos, George Kokotos
ABSTRACT
A tripeptide-like prolinamide-thiourea catalyst with (S)-proline, (1S,2S)-diphenylethylenediamine and (S)-di-tert-butyl aspartate as building blocks provides the products of the reaction between ketones and aromatic aldehydes in high to quantitative yields and high stereoselectivities (up to 99:1 dr and 99% ee). Both the chiral centers of the diamine unit are essential, while the thiourea hydrogen originating from the amine and the amide hydrogen play a predominant role for the catalyst efficiency.
Sigma-Aldrich
L-Prolinamide, 98%