Merck
  • Azidomethyl-ruthenocene: facile synthesis of a useful metallocene derivative and its application in the 'click' labelling of biomolecules.
Azidomethyl-ruthenocene: facile synthesis of a useful metallocene derivative and its application in the 'click' labelling of biomolecules.
Chemical communications (Cambridge, England) (2011-09-22)
Malay Patra, Nils Metzler-Nolte
ABSTRACT
A convenient synthesis of azidomethyl-ruthenocene and its use in the covalent labelling of amino acids, peptides and a peptide nucleic acid (PNA) monomer derivative by Cu(I) catalyzed azide-alkyne coupling (Cu-AAC, "click chemistry") are described.
Sigma-Aldrich
Bis(cyclopentadienyl)ruthenium(II), 97%