Merck
  • Stimulation of medial prefrontal cortex serotonin 2C (5-HT(2C)) receptors attenuates cocaine-seeking behavior.
Stimulation of medial prefrontal cortex serotonin 2C (5-HT(2C)) receptors attenuates cocaine-seeking behavior.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2010-06-04)
Nathan S Pentkowski, Felicia D Duke, Suzanne M Weber, Lara A Pockros, Andrew P Teer, Elizabeth C Hamilton, Kenneth J Thiel, Janet L Neisewander
ABSTRACT
Serotonin 2C receptor (5-HT(2C)R) agonists administered systemically attenuate both cocaine-primed and cue-elicited reinstatement of extinguished cocaine-seeking behavior. To further elucidate the function of these receptors in addiction-like processes, this study examined the effects of microinfusing the 5-HT(2C)R agonist MK212 (0, 10, 30, 100 ng/side/0.2 microl) into the medial prefrontal cortex (mPFC) on cocaine self-administration and reinstatement of extinguished cocaine-seeking behavior. Male Sprague-Dawley rats were trained to self-administer cocaine (0.75 mg/kg, i.v.) paired with light and tone cues. Once responding stabilized, rats received MK212 microinfusions before tests for maintenance of cocaine self-administration. Next, extinction training to reduce cocaine-seeking behavior, defined as responses performed without cocaine reinforcement available, occurred until low extinction baselines were achieved. Rats then received MK212 microinfusions before tests for reinstatement of extinguished cocaine-seeking behavior elicited by cocaine-priming injections (10 mg/kg, i.p.) or response-contingent presentations of the cocaine-associated cues; operant responses during cocaine-primed reinstatement tests produced no consequences. MK212 microinfusions into the prelimbic and infralimbic, but not anterior cingulate, regions of the mPFC dose-dependently attenuated both cocaine-primed and cue-elicited reinstatement of extinguished cocaine-seeking behavior, but did not reliably affect cocaine self-administration. A subsequent experiment showed that the effects of MK212 (100 ng/side/0.2 microl) on reinstatement of extinguished cocaine-seeking behavior were blocked by co-administration of the 5-HT(2C)R antagonist SB242084 (200 ng/side/0.2 microl). MK212 administered alone into the mPFC as a drug prime produced no discernable effects on cocaine-seeking behavior. These findings suggest that stimulation of 5-HT(2C)Rs in the mPFC attenuates the incentive motivational effects produced by sampling cocaine or exposure to drug-paired cues.
Sigma-Aldrich
MK-212 hydrochloride, ≥98% (HPLC)