Merck
  • Chemical and enzymatic N-glycan release comparison for N-glycan profiling of monoclonal antibodies expressed in plants.
Chemical and enzymatic N-glycan release comparison for N-glycan profiling of monoclonal antibodies expressed in plants.
Analytical biochemistry (2010-01-30)
Ada Triguero, Gleysin Cabrera, Louise Royle, David J Harvey, Pauline M Rudd, Raymond A Dwek, Muriel Bardor, Patrice Lerouge, José A Cremata
ABSTRACT
Plants synthesize N-glycans containing the antigenic sugars alpha(1,3)-fucose and beta(1,2)-xylose. Therefore it is important to monitor these N-glycans in monoclonal antibodies produced in plants (plantibodies). We evaluated several techniques to characterize the N-glycosylation of a plantibody produced in tobacco plants with and without the KDEL tetrapeptide endoplasmic reticulum retention signal which should inhibit or drastically reduce the addition of alpha(1,3)-fucose and beta(1,2)-xylose. Ammonium hydroxide/carbonate-based chemical deglycosylation and PNGase A enzymatic release were investigated giving similar 2-aminobenzamide-labeled N-glycan HPLC profiles. The chemical release does not generate peptides which is convenient for MS analysis of unlabeled pool but its main drawback is that it induces degradation of alpha1,3-fucosylated N-glycan reducing terminal sugar. Three analytical methods for N-glycan characterization were evaluated: (i) MALDI-MS of glycopeptides from tryptic digestion; (ii) negative-ion ESI-MS/MS of released N-glycans; (iii) normal-phase HPLC of fluorescently labeled glycans in combination with exoglycosidase sequencing. The MS methods identified the major glycans, but the HPLC method was best for identification and relative quantitation of N-glycans. Negative-mode ESI-MS/MS permitted also the correct identification of the linkage position of the fucose residue linked to the inner core N-acteylglucosamine (GlcNAc) in complex N-glycans.
Sigma-Aldrich
Anthranilamide, ≥98%