- Fatalities associated with clozapine-related constipation and bowel obstruction: a literature review and two case reports.
Fatalities associated with clozapine-related constipation and bowel obstruction: a literature review and two case reports.
Psychosomatics (2009-08-19)
Katharine R Hibbard, Arthur Propst, Daniel E Frank, Jonathan Wyse
PMID19687183
ABSTRACT
Constipation is an exceedingly common side effect of treatment with clozapine. In rare cases, this side effect has resulted in fatal complications. The authors review the literature on fatal complications of clozapine-related constipation and bowel obstruction. The authors provide two new case reports of patients who died of similar causes. There were seven reports of deaths from clozapine-related bowel obstruction in the literature, with the most common mechanisms of death being severe impaction leading either to feculent vomiting or bowel necrosis. The discussion outlines potential mechanisms and management of clozapine-related constipation.
Product Description
Supelco
Flurazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®