Merck
  • Catalysis of 3-pyrrolidinecarboxylic acid and related pyrrolidine derivatives in enantioselective anti-Mannich-type reactions: importance of the 3-acid group on pyrrolidine for stereocontrol.
Catalysis of 3-pyrrolidinecarboxylic acid and related pyrrolidine derivatives in enantioselective anti-Mannich-type reactions: importance of the 3-acid group on pyrrolidine for stereocontrol.
Journal of the American Chemical Society (2008-01-01)
Haile Zhang, Susumu Mitsumori, Naoto Utsumi, Masanori Imai, Noemi Garcia-Delgado, Maria Mifsud, Klaus Albertshofer, Paul Ha-Yeon Cheong, K N Houk, Fujie Tanaka, Carlos F Barbas
ABSTRACT
The development of enantioselective anti-selective Mannich-type reactions of aldehydes and ketones with imines catalyzed by 3-pyrrolidinecarboxylic acid and related pyrrolidine derivatives is reported in detail. Both (3R,5R)-5-methyl-3-pyrrolidinecarboxylic acid and (R)-3-pyrrolidinecarboxylic acid efficiently catalyzed the reactions of aldehydes with alpha-imino esters under mild conditions and afforded anti-Mannich products with high diastereo- and enantioselectivities (anti/syn up to 99:1, up to >99% ee). For the reactions of ketones with alpha-imino esters, (R)-3-pyrrolidinecarboxylic acid was an efficient catalyst (anti/syn up to >99:1, up to 99% ee). Evaluation of a series of pyrrolidine-based catalysts indicated that the acid group at the beta-position of the pyrrolidine ring of the catalyst played an important role in forwarding the carbon-carbon bond formation and in directing anti-selectivity and enantioselectivity.
Sigma-Aldrich
(S)-(+)-Pyrrolidine-3-carboxylic acid, ≥98.0% (NT)