- Guanidinium ylide mediated aziridination: identification of a spiro imidazolidine-oxazolidine intermediate.
Guanidinium ylide mediated aziridination: identification of a spiro imidazolidine-oxazolidine intermediate.
The Journal of organic chemistry (2006-08-12)
Wannaporn Disadee, Tsutomu Ishikawa, Masatoshi Kawahata, Kentaro Yamaguchi
PMID16901150
ABSTRACT
We successfully isolated a spiro imidazolidine-oxazolidine intermediate in the reaction of guanidinium ylide mediated aziridination using alpha-bromocinnamaldehyde. X-ray crystallographic analysis unambiguously revealed that the stereogenic centers of the spiro intermediate were in a trans configuration. The role of the spiro compound as an intermediate in the aziridination reaction was confirmed by observation of its smooth chemical conversion into aziridine products.