Merck
  • [UV spectrophotometric assay of famotidine in combination with picrolonic acid, picrolinate].
[UV spectrophotometric assay of famotidine in combination with picrolonic acid, picrolinate].
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (2006-04-13)
M Apostu, Nela Bibire, V Dorneanu
ABSTRACT
Famotidine, belonging to H2-antagonist group, is a compound containing a thiazolic moiety and it is used in peptic ulcer therapy. This paper debates the possibility of developing a new ultraviolet spectrophotometric assessment by using the reaction between famotidine and picrolonic acid. We carried out our determinations at 362 nm, where the absorbency of famotidine - picrolonic acid complex is maximal, and we have established the optimal reaction conditions. This method was successfully applied for famotidine assay from pharmaceutical dosage forms.