Merck
  • Fluorescence enhancement by hydroperoxides based on a change in the intramolecular charge transfer character of benzofurazan.
Fluorescence enhancement by hydroperoxides based on a change in the intramolecular charge transfer character of benzofurazan.
Chemical communications (Cambridge, England) (2005-03-30)
Maki Onoda, Hidetoshi Tokuyama, Seiichi Uchiyama, Ken-Ichi Mawatari, Tomofumi Santa, Kiyoko Kaneko, Kazuhiro Imai, Kazuya Nakagomi
ABSTRACT
Strong fluorescence signals were observed after the reaction of novel reagents with hydroperoxides.
Sigma-Aldrich
Benzofurazan, 97%