Merck
  • A method to attenuate pneumoperitoneum-induced reductions in splanchnic blood flow.
A method to attenuate pneumoperitoneum-induced reductions in splanchnic blood flow.
Annals of surgery (2005-01-15)
Nishath Athar Ali, W Steve Eubanks, Jonathan S Stamler, Andrew J Gow, Sandhya A Lagoo-Deenadayalan, Leonardo Villegas, Habib E El-Moalem, James D Reynolds
ABSTRACT
To determine if increasing nitric oxide bioactivity by inclusion of ethyl nitrite (ENO) in the insufflation admixture would attenuate pneumoperitoneum-induced decreases in splanchnic perfusion. Organ blood flow is reduced during pneumoperitoneum and can contribute to laparoscopy-associated morbidity and mortality. Previous attempts to control such decreases in flow have been ineffective. Laser-Doppler flow probes were placed on the liver and right kidney of anesthetized pigs. After a baseline recording period, animals were insufflated to a final intraperitoneal pressure of 15 mm Hg. Group one received CO2 (standard practice), whereas group 2 received CO2 plus 100 ppm ENO. Insufflation was maintained for 60 minutes and then the abdomen was manually deflated; monitoring was continued for another 60 minutes. CO2 insufflation (n = 5) cut liver blood flow in half; liver flow remained at this level throughout the postinsufflation period. Inclusion of 100 ppm ENO (n = 6) attenuated both the acute and prolonged blood flow decreases. Statistical modeling of the data showed that, on average, liver blood flow was 14.3 U/min higher in the ENO pigs compared with the CO2 group (P = 0.0454). In contrast, neither treatment significantly altered kidney blood flow (P = 0.6215). The data indicate that ENO can effectively attenuate pneumoperitoneum-induced blood flow decreases within the peritoneal cavity. The result suggests a novel therapeutic method of regulating hemodynamic changes during laparoscopic procedures.
Sigma-Aldrich
Ethyl nitrite solution, 10-20 wt. % in ethanol