Merck
  • [Thermal knowledge and therapies: a comparative view of Portugal (São Pedro do Sul hot springs) and Brazil (Caldas da Imperatriz hot springs)].
[Thermal knowledge and therapies: a comparative view of Portugal (São Pedro do Sul hot springs) and Brazil (Caldas da Imperatriz hot springs)].
Historia, ciencias, saude--Manguinhos (2004-09-29)
Maria Manuel Quintela
ABSTRACT
Based on nineteenth - and twentieth-century texts, the article discusses how medicine legitimized the therapeutic practice of thermalism as so-called 'scientific' knowledge, with the creation of therapeutic establishments and hot-springs resort. My research began in Portugal in 1996, where I produced an ethnography of experiences at the São Pedro do Sul hot springs. My research at Brazil's Caldas da Imperatriz, initiated in August 2001, is still underway.