Merck
  • Differential effect of phosphodiesterase IV inhibitor RP73401 on various inflammatory and immune responses relevent to rheumatoid arthritis.
Differential effect of phosphodiesterase IV inhibitor RP73401 on various inflammatory and immune responses relevent to rheumatoid arthritis.
Pharmacological research (2004-03-05)
Jae Youl Cho, Joon Seok Park, Kyong Up Baik, June Goo Lee, Hyun Pyo Kim, Eun Sook Yoo, Myung Hwan Park
ABSTRACT
Phosphodiesterase (PDE) IV inhibitors have been reported to possess potent anti-inflammatory activities through enhancement of cAMP. In this study, the immunopharmacological effect of PDE IV inhibitor (RP73401) was further carefully evaluated. RP73401 strongly blocked the production of tumor necrosis factor (TNF)-alpha from lipopolysaccharide (LPS)-stimulated murine macrophages (RAW264.7) and human peripheral blood mononuclear cells (PBMC) and LPS-primed mice. RP73401 did not relieve joint inflammation in adjuvant-arthritis (RA) model, whereas the compound attenuated arachidonic acid-induced inflammation. RP73401 displayed weak or no modulatory effects on the activation of macrophage and lymphocytes (assessed by proliferation, nitric oxide (NO) release and cell-cell adhesion, TNF-alpha production upon phorbol 12-myristate 13-acetate (PMA) treatment), and fluorescein-isothiocynate (FITC)-induced ear oedema. Collectively, these data suggest that PDE IV inhibitor RP73401 may differentially modulate various immune responses and these may explain its inability to inhibit adjuvant-induced joint inflammation or FITC-induced ear oedema.
Sigma-Aldrich
Piclamilast, ≥98% (HPLC)