Merck
  • Structure elucidation of a novel funicone-like compound produced by Penicillium pinophilum.
Structure elucidation of a novel funicone-like compound produced by Penicillium pinophilum.
Natural product letters (2002-06-07)
Salvatore de Stefano, Rosario Nicoletti, Salvatore Zambardino, Alfredo Milone
ABSTRACT
A novel metabolite related to the previously identified compound 3-O-methylfunicone (1) was isolated from the fungus Penicillium pinophilum and its structure elucidated by spectral analysis as 3-O-methyl-5, 6-epoxyfunicone (2).