Merck
  • [Method-performance studies of notified analytical method for inabenfide].
[Method-performance studies of notified analytical method for inabenfide].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2002-01-31)
K Sasaki, T Tatsuno, M Nakamura, A Anazawa, T Imazawa, O Utsunomiya, S Terasawa, H Osada, M Kaneko, M Toyoda
ABSTRACT
Method-performance studies were conducted for the notified revised analytical method of inabenfide in unpolished rice. Six laboratories analyzed unpolished rice spiked with 0.05 microgram/g of inabenfide in replicate. Mean recovery from rice was 85.0%. Repeatability relative standard deviation value was 4.2% and reproducibility relative standard deviation value was 8.1%. The detection limits were 0.002-0.01 microgram/g.