Merck
Photoinduced reactions of 1-acetylisatin with phenylacetylenes.
Organic letters (2000-09-16)
J Xue, Y Zhang, X I Wang, H K Fun, J H Xu
ABSTRACT
[reaction: see text]Photoinduced reactions of 1-acetylisatin 1 with diphenylacetylenes 2a-2c afforded the corresponding 3-methylene-2-indolones 4a-4e and 5a-5e in 80-90% yields via a spirooxetene intermediate. Similar irradiation of 1 with phenylacetylene, on the other hand, resulted in efficient formation of two diastereoisomeric dispiro[3H-indole-3,2'-furan-3',3"-(3H)-indole]-2,5',2"(2H,5'H,2"H )triones 6 and 7 via a reaction sequence with initial formation of the spirooxetene intermediate. The regioselectivity in the photocycloaddition of 1 with phenylacetylene and the reaction mechanism for the formation of 6 and 7 are discussed.