Merck
Convenient synthesis of L-proline benzyl ester.
Bioorganic & medicinal chemistry letters (1999-11-24)
A Córdova, N N Reed, J A Ashley, K D Janda
ABSTRACT
Mesylates or tosylates of delta-hydroxy-L-norvaline esters spontaneously afford L-proline esters upon exposure to aqueous buffer in near quantitative yield. This novel reaction has led to the development of a simple route to optically active proline esters.
Sigma-Aldrich
L-Proline benzyl ester hydrochloride, 98%
Pricing and availability is not currently available.