Merck
Triterpenoidal saponins from the bark of Kalopanax pictum var. typicum.
Archives of pharmacal research (1991-03-01)
S H Cho, D R Hahn
ABSTRACT
One new triterpenoidal saponin, saponin F(2) has been isolated from the bark of Kalopanax pictum Nakai var. typicum (Araliaceae), together with one known saponin, kizuta saponin K12 (1). On the basis of chemico-spectral evidences, the structure of 2 has been elucidated to be 3-O-beta-D-xylopyranosyl(1-->3)-alpha- L-rhamnopyranosyl(1-->2)-alpha- L-arabinopyranosyl-23-hydroxyolean-12-en-28-O-alpha-L- rhamnopyranosyl(1-->4)-beta-D-glucopyranosyl(1-->6)-beta-D-glucopyranosy l ester.
Product Description
Supelco
Hederacoside C, analytical standard
Pricing and availability is not currently available.
Hederacoside C, primary reference standard
Pricing and availability is not currently available.