Merck
  • Synthesis of 1-[11C]methylpiperidin-4-yl propionate ([11C]PMP) for in vivo measurements of acetylcholinesterase activity.
Synthesis of 1-[11C]methylpiperidin-4-yl propionate ([11C]PMP) for in vivo measurements of acetylcholinesterase activity.
Nuclear medicine and biology (1998-12-24)
S E Snyder, L Tluczek, D M Jewett, T B Nguyen, D E Kuhl, M R Kilbourn
ABSTRACT
Synthesis of 1-[11C]methylpiperidin-4-yl propionate ([11C]PMP), an in vivo substrate for acetylcholinesterase, is reported. An improved preparation of 4-piperidinyl propionate (PHP), the immediate precursor for radiolabeling, was accomplished in three steps from 4-hydroxypiperidine by (a) protection of the amine as the benzyl carbamate, (b) acylation with propionyl chloride, and (c) deprotection of the carbamate by catalytic hydrogenation. The final product was obtained in an overall 82% yield. Reaction of the free base form of PHP with [11C]methyl trifluoromethanesulfonate at room temperature in N,N-dimethylformamide, followed by high performance liquid chromatography (HPLC) purification, provided [11C]PMP in 57% radiochemical yield, > 99% radiochemical purity, and > 1500 Ci/mmol at the end of synthesis. The total synthesis time from end-of-bombardment was 35 min. [11C]PMP can thus be reliably prepared for routine clinical studies of acetylcholinesterase in human brain using positron emission tomography.
Product Description
Supelco
Methyl trifluoromethanesulfonate, for GC derivatization, LiChropur, 98.0%
Sigma-Aldrich
Methyl trifluoromethanesulfonate, ≥98%