Merck
  • Serratia marcescens chitobiase is a retaining glycosidase utilizing substrate acetamido group participation.
Serratia marcescens chitobiase is a retaining glycosidase utilizing substrate acetamido group participation.
The Biochemical journal (1998-02-07)
S Drouillard, S Armand, G J Davies, C E Vorgias, B Henrissat
ABSTRACT
The stereochemistry of the reaction catalysed by Serratia marcescens chitobiase was determined by HPLC separation of the anomers of N-acetylglucosamine produced during the hydrolysis of p-nitrophenyl N-acetyl-beta-d-glucosaminide (PNP-GlcNAc). In the early stages of the reaction, the beta-anomer was found to prevail, whereas the alpha-anomer dominated at mutarotation equilibrium. This established that chitobiase hydrolyses glycosidic bonds with overall retention of the anomeric configuration. Chitobiase-catalysed hydrolysis of PNP-GlcNAc was competitively inhibited by a series of chito-oligosaccharides (degree of polymerization 2-5) that were selectively de-N-acetylated at their non-reducing end. The results are in accord with the participation of the acetamido group at C-2 of the substrate in the catalytic mechanism of chitobiase and related enzymes.
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitrophenyl N-acetyl-α-D-glucosaminide, ≥98%
Pricing and availability is not currently available.