Merck
  • Radical scavenging activity and cytotoxicity of active quinic acid derivatives from Scorzonera divaricata roots.
Radical scavenging activity and cytotoxicity of active quinic acid derivatives from Scorzonera divaricata roots.
Food chemistry (2013-02-16)
Yong-Jin Yang, Xu Liu, Hong-Ru Wu, Xiao-Feng He, Yu-Rong Bi, Ying Zhu, Zhen-Ling Liu
ABSTRACT
Five new quinic acid derivatives and two known 3-O-feruloylquinic acids were isolated from the roots of Scorzonera divaricata Turcz. The new compounds were elucidated as (-)-1,4-di-O-feruloyl-3-O-dihydrocaffeoylquinic acid, (-)-1-O-feruloyl-4-O- dihydrocaffeoylquinic acid, (-)-3,5-di-O-feruloylquinic acid, (-)-1-O-feruloyl-3-O-dihydro- caffeoylquinic acid, and (-)-1-O-feruloyl-5-O-dihydrocaffeoylquinic acid based on extensive spectroscopic studies, including one- and two-dimensional NMR, HRESIMS, UV, and IR results. Five compounds were assessed for antioxidant activity by ABTS and DPPH radical-scavenging assays and for their cytotoxicity against HL-60 and Hep-G2 cell lines by the MTT assay. Three quinic acid derivatives exhibited strong antioxidant activity, with IC(50) values of 3.95, 5.87, and 7.45 μg/mL against ABTS(+) and 11.7, 13.6, and 50.1 μg/mL against DPPH(). (-)-1,4-Di-O-feruloyl-3-O-dihydrocaffeoylquinic acid also exhibited moderate activity against Hep-G2 cell lines with an IC(50) value of 14.6 μg/mL.
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Supelco
(−) Quinic acid, analytical standard