Merck
Dissociative electron attachment to triflates.
The Journal of chemical physics (2011-12-14)
Sylwia Ptasińska, David Gschliesser, Peter Bartl, Ireneusz Janik, Paul Scheier, Stephan Denifl
ABSTRACT
Gas phase studies of dissociative electron attachment to simple alkyl (CF(3)SO(3)CH(3)) and aryl (C(6)H(5)SO(3)CF(3) and CF(3)SO(3)C(6)H(4)CH(3)) triflates, model molecules of nonionic photoacid generators for modern lithographic applications, were performed. The fragmentation pathways under electron impact below 10 eV were identified by means of crossed electron-molecular beam mass spectrometry. Major dissociation channels involved C-O, S-O, or C-S bond scissions in the triflate moiety leading to the formation of triflate (OTf(-)), triflyl (Tf(-)), or sulfonate (RSO(3)(-)) anions, respectively. A resonance leading to C-O bond breakage and OTf(-) formation in alkyl triflates occurred at electron energies about 0.5 eV lower than the corresponding resonance in aryl triflates. A resonance leading to S-O bond breakage and Tf(-) formation in aryl triflates occurred surprisingly at the same electron energies as C-O bond breakage. In case of alkyl triflates S-O bond breakage required 1.4 eV higher electron energies to occur and proceeded with substantially lower yields than in aryl triflates. C-S bond scission occurred for all presently studied triflates at energies close to 3 eV.
Product Description
Supelco
Methyl trifluoromethanesulfonate, for GC derivatization, LiChropur, 98.0%
Sigma-Aldrich
Methyl trifluoromethanesulfonate, ≥98%