- Facile preparation of alpha-aryl nitriles by direct cyanation of alcohols with TMSCN under the catalysis of InX3.
Facile preparation of alpha-aryl nitriles by direct cyanation of alcohols with TMSCN under the catalysis of InX3.
Organic letters (2008-09-24)
Gang Chen, Zheng Wang, Jiang Wu, Kuiling Ding
PMID18808133
ABSTRACT
A convenient and efficient synthesis of alpha-aryl nitriles was developed by direct cyanation of alcohols with TMSCN under the catalysis of Lewis acid. Using 5-10 mol % of InBr3 as the catalyst, a variety of benzylic alcohols can be converted to the corresponding nitriles in 5-30 min with yields of 46-99%.