Merck
  • Fluorescent detection of peptides and amino acids for capillary electrophoresis via on-line derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole.
Fluorescent detection of peptides and amino acids for capillary electrophoresis via on-line derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole.
Analytical and bioanalytical chemistry (2006-09-06)
Hongyi Zhang, Isabelle Le Potier, Claire Smadja, Jiyou Zhang, Myriam Taverna
ABSTRACT
An in-capillary derivatization of amino acids and peptides with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F) was developed for their subsequent capillary electrophoretic analysis with laser-induced fluorescence detection (lambda (ex)=488 nm). The in-capillary derivatization was achieved in zone-passing mode by introducing successive plugs of sample and NBD-F into a fused silica capillary previously equilibrated with an alkaline borate buffer. To prevent NBD-F hydrolysis and to achieve a reliable derivatization, NBD-F was prepared daily in absolute ethanol and a plug of absolute ethanol was introduced between the sample and NBD-F reagent plugs. Various parameters influencing the derivatization efficiency were investigated and the optimum conditions were as follows: background electrolyte (BGE), 20 mM borate buffer (pH 8.8); introduction time, 4 s for sample and 2 s for NBD-F; molar ratio of NBD-F/sample, above 215; temperature, 45 degrees C for amino acids and 35 degrees C for peptides; applied voltage, +15 kV. The validation of the in-capillary derivatization method under optimal conditions showed a good linearity between the heights of the derivative peaks and the concentrations of the amino acids. The intra-day relative standard deviations of the migration times and the peak heights were less than 1.3% and 4.6%, respectively. The efficient derivatization and separation of a mixture of valine, alanine, glutamic acid and aspartic acid were achieved using this technique. Peptides such as buccaline and beta-protein fragment 1-42 could also be derivatized using the developed in-capillary derivatization procedure.
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, ≥98% (elemental analysis)