Merck
  • Sample metallization for performance improvement in desorption/ionization of kilodalton molecules: quantitative evaluation, imaging secondary ion MS, and laser ablation.
Sample metallization for performance improvement in desorption/ionization of kilodalton molecules: quantitative evaluation, imaging secondary ion MS, and laser ablation.
Analytical chemistry (2003-12-13)
A Delcorte, J Bour, F Aubriet, J-F Muller, P Bertrand
ABSTRACT
The metallization procedure, proposed recently for signal improvement in organic secondary ion mass spectrometry (SIMS) (Delcorte, A.; Médard, N.; Bertrand, P. Anal.Chem. 2002, 74, 4955)., has been thoroughly tested for a set of kilodalton molecules bearing various functional groups: Irganox 1010, polystyrene, polyalanine, and copper phthalocyanine. In addition to gold, we evaluate the effect of silver evaporation as a sample treatment prior to static SIMS analysis. Ion yields, damage cross sections, and emission efficiencies are compared for Ag- and Au-metallized molecular films, pristine coatings on silicon, and submonolayers of the same molecules adsorbed on silver and gold. The results are sample-dependent but as an example, the yield enhancement calculated for metallized Irganox films with respect to untreated coatings is larger than 2 orders of magnitude for the quasimolecular ion and a factor of 1-10 for characteristic fragments. Insights into the emission processes of quasimolecular ions from metallized surfaces are deduced from kinetic energy distribution measurements. The advantage of the method for imaging SIMS applications is illustrated by the study of a nonuniform coating of polystyrene oligomers on a 100-microm polypropylene film. The evaporated metal eliminates sample charging and allows us to obtain enhanced quality images of characteristic fragment ions as well as reasonably contrasted chemical mappings for cationized PS oligomers and large PP chain segments. Finally, we report on the benefit of using metal evaporation as a sample preparation procedure for laser ablation mass spectrometry. Our results show that the fingerprint spectra of Au-covered polystyrene, polypropylene, and Irganox films can be readily obtained under 337-nm irradiation, a wavelength for which the absorption of polyolefins is low. This is probably because the gold clusters embedded in the sample surface absorb and transfer the photon energy to the surrounding organic medium.
Sigma-Aldrich
Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), 98%
Plastic additive 09, European Pharmacopoeia (EP) Reference Standard