Merck
  • Detection of the novel metabolite ethylphenidate after methylphenidate overdose with alcohol coingestion.
Detection of the novel metabolite ethylphenidate after methylphenidate overdose with alcohol coingestion.
Journal of clinical psychopharmacology (1999-08-10)
J S Markowitz, B K Logan, F Diamond, K S Patrick
ABSTRACT
Methylphenidate is the most commonly prescribed psychostimulant in clinical use today. Known methylphenidate metabolites include ritalinic acid, corresponding lactams, and p-hydroxymethylphenidate. Recent in vitro work using rat liver preparations has indicated that the methylphenidate ethyl ester, ethylphenidate, is formed upon incubation with ethanol. This report describes the first detection of ethylphenidate in human blood and liver samples obtained from two suicide victims who had overdosed on methylphenidate and coingested ethanol. Amounts of ethylphenidate detected in whole blood specimens in these two cases (8 ng/mL and 1 ng/mL, respectively) were small relative to methylphenidate and ritalinic acid concentrations. Nonetheless, given the high likelihood that methylphenidate and ethanol coingestion frequently occurs, the detection of ethylphenidate in humans warrants further investigation into the extent of its formation as well as into any associated toxicity in nonoverdose situations.
Product Description
Supelco
Ritalinic acid hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ritalinic acid, 99% (CP)