Merck
  • Influence of growth mode and sucrose on susceptibility of Streptococcus sanguis to amine fluorides and amine fluoride-inorganic fluoride combinations.
Influence of growth mode and sucrose on susceptibility of Streptococcus sanguis to amine fluorides and amine fluoride-inorganic fluoride combinations.
Applied and environmental microbiology (1998-09-03)
J V Embleton, H N Newman, M Wilson
ABSTRACT
This study evaluated the susceptibility to amine fluorides (AmFs) of planktonic and biofilm cultures of Streptococcus sanguis grown with and without sucrose. Cultures were incubated with AmFs (250 mg of fluoride liter-1) for 1 min. The susceptibility of biofilms was less than that of the planktonic form and was further decreased by growth in the presence of sucrose.
Sigma-Aldrich
Zinc fluoride, powder, 99%