Merck
  • Simulation of in situ subsurface biodegradation of polychlorophenols in air-lift percolators.
Simulation of in situ subsurface biodegradation of polychlorophenols in air-lift percolators.
Applied microbiology and biotechnology (1998-07-31)
J H Langwaldt, M K Männistö, R Wichmann, J A Puhakka
ABSTRACT
Air-lift percolator experiments simulated in situ subsurface degradation of 2,4,6-trichlorophenol, 2,3,4,6-tetrachlorophenol and pentachlorophenol, in mixtures and individually, by indigenous microorganisms from a chlorophenol-contaminated aquifer. Inoculation with a chlorophenol(CP)-degrading gram-positive isolate from the CP-contaminated groundwater did not significantly increase CP degradation rates. Feed concentrations of up to 55 mg CP l-1 were degraded. Stable CP degradation was maintained for over 6 months. CP degradation rates up to 54.3 mg l-1 day-1 and effluent concentrations below 40 micrograms CP l-1 were achieved. CP were mineralized as shown by CP reduction, dissolved organic carbon removal and release of inorganic chloride.
Product Description
Supelco
2,3,4,6-Tetrachlorophenol, analytical standard
Supelco
2,3,4,6-Tetrachlorophenol solution, certified reference material, 5000 μg/mL in methanol