Merck
  • Disposition of human drug preparations in the horse. V. Orally administered oxprenolol.
Disposition of human drug preparations in the horse. V. Orally administered oxprenolol.
Biomedical chromatography : BMC (1996-07-01)
F T Delbeke
ABSTRACT
Urinary concentrations of the beta-antagonist oxprenolol and some of its major human metabolites were determined following oral administration of a dose of 160 mg to five fasted horses. Quantitation was performed by gas chromatography-mass spectrometry (GC-MS) in the selected ion mode (SIM) by monitoring ion m/z 466 of the heptafluorobutyric derivatives. As early as 2 h after dosage oxprenolol could be detected in hydrolysed urine and remained detectable up to 24 h. Maximum urinary concentrations and excretion rates were obtained between 2 and 12 h. After 12 h only 2.8% of the administered dose was excreted as conjugates of oxprenolol and major human metabolites including 4-OH-oxprenolol and 5-OH-oxprenolol. These metabolites were detectable up to 48 h.
Sigma-Aldrich
Heptafluorobutyric anhydride, ≥98%
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%