Merck
  • In vitro and in vivo metabolism of the antianxiolytic agent fenobam in the rat.
In vitro and in vivo metabolism of the antianxiolytic agent fenobam in the rat.
Journal of pharmaceutical sciences (1995-02-01)
W N Wu, L A McKown, P J O'Neill
ABSTRACT
Fenobam [(Fn); N-(3-chlorophenyl)-N-(4,5-dihydro-1-methyl-4-oxo-1H-imidazole-2-yl)urea] sulfate is a novel agent with potent anxiolytic activity in rats. [14C]Fn sulfate was administered as an oral solution (250 mg/kg) to male Wistar rats, and 52% of the administered dose was excreted in urine (0-5 days). In vitro metabolism of Fn was studied by incubating [14C]Fn with rat hepatic 9000 x g supernatant preparations. Unchanged Fn and a total of six metabolites were isolated, quantified, and identified from the urine and liver 9000 x g supernatant samples by column chromatography; TLC; UV, IR, and NMR spectroscopy; MS; and comparison with synthetic samples. Four metabolic pathways for Fn are proposed: (1) hydroxylation at the phenyl ring to form 4-hydroxyphenyl-Fn, a major pathway in vivo (12% of the sample radioactivity) but a minor pathway in vitro (4% of the sample radioactivity); (2) hydroxylation at the creatinine ring to form 5-hydroxy-Fn (19%) of the sample radioactivity), a dominant pathway in vitro but not in vivo; (3) oxidative cleavage at the creatinine ring (loss of a ketene unit), a minor pathway for Fn but an important pathway for 4-hydroxyphenyl-Fn in vivo; and (4) N-demethylation, a minor pathway for Fn in vivo.
Sigma-Aldrich
Fenobam, ≥98% (HPLC), solid