Merck
  • Inositol trisphosphate-dependent and -independent Ca2+ mobilization pathways at the vacuolar membrane of Candida albicans.
Inositol trisphosphate-dependent and -independent Ca2+ mobilization pathways at the vacuolar membrane of Candida albicans.
The Journal of biological chemistry (1995-03-31)
C M Calvert, D Sanders
ABSTRACT
Vacuolar membrane vesicles were isolated from Candida albicans protoplasts, and marker enzyme assays were employed to identify the membranes as vacuolar in origin. The mechanisms of Ca2+ uptake and Ca2+ release at the vacuolar membrane were investigated. Ca2+ accumulation by vacuolar membrane vesicles can be generated via H+/Ca2+ antiport. The inside-acid pH is in turn generated by a vacuolar-type H(+)-ATPase, as demonstrated by the sensitivity of Ca2+ uptake to ionophores and the vacuolar H(+)-ATPase inhibitor bafilomycin A1. Vacuolar membrane vesicles exhibit two Ca2+ release pathways: one induced by inositol 1,4,5-trisphosphate (InsP3) and the other by inside-positive voltage. These two pathways are distinct with respect to the amount of Ca2+ released, the nature of response to successive stimuli, and their respective pharmacological profiles. The InsP3-gated pathway exhibits a K0.5 for InsP3 of 2.4 microM but is not activated by inositol 4,5-bisphosphate or inositol 1,3,4,5-tetrakisphosphate at concentrations up to 50 microM. Ca2+ release by InsP3 is blocked partially by low molecular weight heparin. Ca2+ released by the voltage-sensitive pathway occurs at membrane potentials estimated to be over a physiological range from 0 to 80 mV. The voltage-sensitive Ca2+ release pathway can be blocked by lanthanide ions and organic channel blockers such as ruthenium red and verapamil. Furthermore, the voltage-sensitive Ca2+ release pathway exhibits Ca(2+)-induced Ca2+ release. These findings are discussed in relation to the mechanism of Ca(2+)-mediated cellular signaling in C. albicans and other fungi.
Sigma-Aldrich
Methyltriphenylphosphonium bromide, 98%
Sigma-Aldrich
Bafilomycin A1, InSolution, ≥90%, A macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase